Assay Details
Target Gene Details
Entrez Gene ID: | 55531 |
Gene Name: | ELMO domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.11:107591091-107666779 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMOD1 | NM_001130037.1 | NP_001123509.1 | ||
| NM_001308018.1 | NP_001294947.1 | |||
| NM_018712.3 | NP_061182.3 | |||
| XM_017017994.1 | XP_016873483.1 | |||
| XM_017017995.1 | XP_016873484.1 | |||
| AK054794.1 | ||||
| AK297351.1 | ||||
| AK303689.1 | ||||
| AL359601.1 | CAB94879.1 | |||
| BC028725.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556377 | Chr.11:107602007 - 107695859 on Build GRCh38 | Gain |
|
| nsv476 | Chr.11:107646652 - 107680407 on Build GRCh38 | Insertion |
|
| esv2422201 | Chr.11:107608636 - 107685099 on Build GRCh38 | Duplication |
|
| dgv1274n100 | Chr.11:107597955 - 108083284 on Build GRCh38 | Gain |
|
| nsv1052198 | Chr.11:107639192 - 107813645 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map