Assay Details
Target Gene Details
Entrez Gene ID: | 6249 |
Gene Name: | CAP-Gly domain containing linker protein 1 |
Gene Aliases: |
CLIP, CLIP-170, CLIP170, CYLN1, RSN |
Location: |
Chr.12:122271432-122422956 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIP1 | NM_001247997.1 | 26 | 5009 | NP_001234926.1 |
| NM_002956.2 | 25 | 4976 | NP_002947.1 | |
| NM_198240.1 | 24 | 4871 | NP_937883.1 | |
| XM_005253593.2 | 27 | 5078 | XP_005253650.1 | |
| XM_006719552.3 | 26 | 5071 | XP_006719615.1 | |
| XM_017019786.1 | 25 | 5140 | XP_016875275.1 | |
| XM_017019787.1 | 26 | 4961 | XP_016875276.1 | |
| XM_017019788.1 | 25 | 4966 | XP_016875277.1 | |
| XM_017019789.1 | 25 | 5125 | XP_016875278.1 | |
| XM_017019790.1 | 24 | 4861 | XP_016875279.1 | |
| XM_017019791.1 | 25 | 4954 | XP_016875280.1 | |
| XM_017019792.1 | 24 | 4849 | XP_016875281.1 | |
| AB209115.1 | 22 | 3735 | BAD92352.1 | |
| AK126990.1 | 16 | 2665 | ||
| M97501.1 | 24 | 4871 | AAA35693.1 | |
| X64838.1 | 25 | 4953 | CAA46050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630973 | Chr.12:122263863 - 122274137 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map