Assay Details
Target Gene Details
Entrez Gene ID: | 51175 |
Gene Name: | tubulin epsilon 1 |
Gene Aliases: |
TUBE, dJ142L7.2 |
Location: |
Chr.6:112070657-112087548 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBE1 | NM_016262.4 | NP_057346.1 | ||
| XM_011535875.2 | XP_011534177.1 | |||
| XM_017010918.1 | XP_016866407.1 | |||
| AF201334.1 | AAF09585.1 | |||
| AK026242.1 | ||||
| AK126191.1 | 1 | 2358 | ||
| BC025405.2 | AAH25405.1 | |||
| BC031101.1 | AAH31101.1 |
More Information
Set Membership: |
|

Genomic Map