Assay Details
Target Gene Details
Entrez Gene ID: | 729920 |
Gene Name: | isoprenoid synthase domain containing |
Gene Aliases: |
MDDGA7, MDDGC7, Nip, hCG_1745121 |
Location: |
Chr.7:16087527-16422562 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISPD | NM_001101417.3 | NP_001094887.1 | ||
| NM_001101426.3 | NP_001094896.1 | |||
| XM_006715770.3 | XP_006715833.1 | |||
| XM_011515499.2 | XP_011513801.1 | |||
| XM_011515500.2 | XP_011513802.1 | |||
| XM_011515508.2 | XP_011513810.1 | |||
| XM_011515509.2 | XP_011513811.1 | |||
| XM_017012575.1 | XP_016868064.1 | |||
| XM_017012577.1 | XP_016868066.1 | |||
| XM_017012578.1 | XP_016868067.1 | |||
| BE092109.1 | ||||
| DY654823.1 |
Target Gene Details
Entrez Gene ID: | 100506025 |
Gene Name: | ISPD antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:16210486-16270604 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISPD-AS1 | NR_038946.1 | |||
| NR_038947.1 | ||||
| BX115602.1 | ||||
| DB462496.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| esv2759514 | Chr.7:16196537 - 16487765 on Build GRCh38 | Loss |
|
| nsv470133 | Chr.7:16230007 - 16272431 on Build GRCh38 | Loss |
|
| esv34298 | Chr.7:16212129 - 16284560 on Build GRCh38 | Loss |
|
| esv2659119 | Chr.7:16207976 - 16291337 on Build GRCh38 | Deletion |
|
| esv3612387 | Chr.7:16208042 - 16291349 on Build GRCh38 | Loss |
|
| nsv1022814 | Chr.7:16222675 - 16300363 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map