Assay Details
Target Gene Details
Entrez Gene ID: | 387644 |
Gene Name: | long intergenic non-protein coding RNA 202-1 |
Gene Aliases: |
C10orf51, LINC00202, NCRNA00202, bB27G4.1 |
Location: |
Chr.10:26931206-26942001 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00202-1 | NR_026795.1 | 5 | 3440 | |
| AK097405.1 | 4 | 1607 | ||
| AK128027.1 | 4 | 5807 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6154 | Chr.10:26923577 - 26937091 on Build GRCh38 | Insertion |
|
| nsv831812 | Chr.10:26772577 - 26945338 on Build GRCh38 | Gain |
|
| esv3415799 | Chr.10:26640952 - 26944612 on Build GRCh38 | Duplication |
|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| nsv1036080 | Chr.10:26926900 - 26941064 on Build GRCh38 | Gain |
|
| esv2759740 | Chr.10:26872003 - 26941064 on Build GRCh38 | Gain+Loss |
|

Genomic Map