Assay Details
Target Gene Details
Entrez Gene ID: | 137886 |
Gene Name: | UBX domain protein 2B |
Gene Aliases: |
p37 |
Location: |
Chr.8:58411264-58451508 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN2B | NM_001077619.1 | NP_001071087.1 | ||
| XM_005251165.2 | XP_005251222.1 | |||
| XM_005251166.1 | XP_005251223.1 | |||
| AK126300.1 | ||||
| AL831990.2 | ||||
| AL833075.2 | ||||
| BC113645.1 | ||||
| BX647876.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759619 | Chr.8:58392731 - 58429772 on Build GRCh38 | Gain |
|
| nsv971656 | Chr.8:58419267 - 58426647 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs71557721] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map