Assay Details
Target Gene Details
Entrez Gene ID: | 440050 |
Gene Name: | keratin associated protein 5-7 |
Gene Aliases: |
KRTAP5-3, KRTAP5.7 |
Location: |
Chr.11:71527267-71528164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP5-7 | NM_001012503.1 | 1 | 624 | NP_001012521.1 |
| AB126076.1 | 1 | 624 | BAD20203.1 | |
| AJ628243.1 | 1 | 210 | CAF31636.1 | |
| BC136967.1 | 1 | 613 | ||
| BC136968.1 | 1 | 613 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2674144 | Chr.11:71503976 - 71537547 on Build GRCh38 | Deletion |
|
| esv3626778 | Chr.11:71503993 - 71537515 on Build GRCh38 | Loss |
|
| esv3626780 | Chr.11:71521100 - 71539367 on Build GRCh38 | Gain |
|
| esv2744750 | Chr.11:71526540 - 71564429 on Build GRCh38 | Deletion |
|
| nsv555402 | Chr.11:71489274 - 71578043 on Build GRCh38 | Gain |
|
| esv3626781 | Chr.11:71526772 - 71565036 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map