Assay Details
Target Gene Details
Entrez Gene ID: | 4115 |
Gene Name: | MAGE family member B4 |
Gene Aliases: |
CT3.6 |
Location: |
Chr.X:30241940-30244193 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB4 | NM_002367.3 | 1 | 925 | NP_002358.1 |
| AK313769.1 | 1 | 865 | ||
| BC032852.2 | 1 | 813 | AAH32852.1 | |
| CR541779.1 | 1 | 729 | CAG46578.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map