Assay Details
Target Gene Details
Entrez Gene ID: | 1270 |
Gene Name: | ciliary neurotrophic factor |
Gene Aliases: |
HCNTF |
Location: |
Chr.11:58622673-58625733 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTF | NM_000614.3 | 2 | 1837 | NP_000605.1 |
| BC068030.1 | 2 | 1832 | AAH68030.1 | |
| BC069167.1 | 2 | 1837 | AAH69167.1 | |
| DB341875.1 | 1 | 55 |
Target Gene Details
Entrez Gene ID: | 386607 |
Gene Name: | ZFP91-CNTF readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:58579114-58625733 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP91-CNTF | NR_024091.1 | 13 | 3487 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159883 | Chr.11:58621846 - 58666718 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map