Assay Details
Target Gene Details
Entrez Gene ID: | 9724 |
Gene Name: | UTP14, small subunit processome component homolog C (S. cerevisiae) |
Gene Aliases: |
2700066J21Rik, KIAA0266, UTP14B |
Location: |
Chr.13:52024691-52033600 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTP14C | NM_021645.5 | 2 | 1944 | NP_067677.4 |
| BC089407.1 | 1 | 1353 | AAH89407.1 | |
| BC122536.1 | 2 | 1721 | ||
| D87455.1 | 2 | 1944 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
| dgv1652n100 | Chr.13:51747428 - 52069788 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map