Assay Details
Target Gene Details
Entrez Gene ID: | 404636 |
Gene Name: | family with sequence similarity 45 member A |
Gene Aliases: |
- |
Location: |
Chr.10:119104065-119137984 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM45A | NM_001303111.1 | NP_001290040.1 | ||
| NM_001303112.1 | NP_001290041.1 | |||
| NM_001303113.1 | NP_001290042.1 | |||
| NM_207009.3 | NP_996892.1 | |||
| NR_130122.1 | ||||
| XM_006717849.3 | XP_006717912.1 | |||
| XM_006717850.3 | XP_006717913.1 | |||
| XM_006717851.3 | XP_006717914.1 | |||
| XM_006717852.3 | XP_006717915.1 | |||
| XM_017016261.1 | XP_016871750.1 | |||
| XM_017016262.1 | XP_016871751.1 | |||
| XM_017016263.1 | XP_016871752.1 | |||
| XM_017016264.1 | XP_016871753.1 | |||
| XM_017016265.1 | XP_016871754.1 | |||
| XM_017016266.1 | XP_016871755.1 | |||
| XM_017016267.1 | XP_016871756.1 | |||
| AK000031.1 | BAA90894.1 | |||
| AK025354.1 | 1 | 65 | ||
| AK027029.1 | ||||
| AK293136.1 | ||||
| AK293365.1 | ||||
| AK297633.1 | ||||
| AK297971.1 | ||||
| BC022271.1 | AAH22271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552214 | Chr.10:119100409 - 119209705 on Build GRCh38 | Loss |
|
| nsv1041093 | Chr.10:118689033 - 119198373 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map