Assay Details
Target Gene Details
Entrez Gene ID: | 84616 |
Gene Name: | keratin associated protein 4-4 |
Gene Aliases: |
KAP4.13, KAP4.4, KRTAP4-13, KRTAP4.13, KRTAP4.4 |
Location: |
Chr.17:41159653-41160731 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP4-4 | NM_032524.1 | 1 | 771 | NP_115913.1 |
| AJ296168.1 | 2 | 594 | CAC27563.1 | |
| AJ406936.1 | 1 | 771 | CAC27575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833446 | Chr.17:41059613 - 41247007 on Build GRCh38 | Gain |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|

Genomic Map