Assay Details
Target Gene Details
Entrez Gene ID: | 100288079 |
Gene Name: | microtubule associated protein 1 light chain 3 beta pseudogene |
Gene Aliases: |
- |
Location: |
Chr.1:185323846-185335039 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GS1-279B7.1 | NR_038424.1 | |||
| AF508905.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758981 | Chr.1:185324065 - 185376639 on Build GRCh38 | Loss |
|
| nsv548398 | Chr.1:185167326 - 186033962 on Build GRCh38 | Gain |
|

Genomic Map