Assay Details
Target Gene Details
Entrez Gene ID: | 7062 |
Gene Name: | trichohyalin |
Gene Aliases: |
THH, THL, TRHY |
Location: |
Chr.1:152106317-152115454 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCHH | NM_007113.3 | 3 | 4028 | NP_009044.2 |
| BC131826.1 | 2 | 3579 | ||
| BC131827.1 | 2 | 3579 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
| nsv438037 | Chr.1:152107513 - 152111563 on Build GRCh38 | Loss |
|
| nsv952187 | Chr.1:152107125 - 152113624 on Build GRCh38 | Deletion |
|
| nsv2866 | Chr.1:152088370 - 152120918 on Build GRCh38 | Insertion |
|
| nsv547853 | Chr.1:152036195 - 152264284 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map