Assay Details
Target Gene Details
Entrez Gene ID: | 100129434 |
Gene Name: | uncharacterized LOC100129434 |
Gene Aliases: |
- |
Location: |
Chr.2:56173534-56185770 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129434 | NR_125368.1 | 3 | 443 | |
| AK096196.1 | 3 | 655 | BAC04724.1 | |
| AK295617.1 | 4 | 1220 | ||
| AK311113.1 | 3 | 443 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575244 | Chr.2:55522427 - 56940881 on Build GRCh38 | Gain |
|
| esv2759051 | Chr.2:55951304 - 56245259 on Build GRCh38 | Gain |
|
| esv2762648 | Chr.2:56159521 - 56195313 on Build GRCh38 | Gain |
|
| esv3892459 | Chr.2:56143744 - 56411721 on Build GRCh38 | Loss |
|

Genomic Map