Assay Details
Target Gene Details
Entrez Gene ID: | 101929710 |
Gene Name: | uncharacterized LOC101929710 |
Gene Aliases: |
- |
Location: |
Chr.5:95962001-96631085 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929710 | NR_130776.1 | |||
| DA871062.1 | ||||
| DB197007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823146 | Chr.5:95965208 - 96117853 on Build GRCh38 | Gain |
|
| dgv1113e214 | Chr.5:95963962 - 96119963 on Build GRCh38 | Gain |
|
| esv2759355 | Chr.5:95896002 - 96204729 on Build GRCh38 | Gain |
|
| esv34203 | Chr.5:95975281 - 96116620 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75828864] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map