Assay Details
Target Gene Details
Entrez Gene ID: | 3619 |
Gene Name: | inner centromere protein |
Gene Aliases: |
- |
Location: |
Chr.11:62123973-62155474 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INCENP | NM_001040694.1 | NP_001035784.1 | ||
| NM_020238.2 | NP_064623.2 | |||
| XM_006718533.2 | XP_006718596.1 | |||
| XM_011544995.2 | XP_011543297.1 | |||
| XM_011544996.2 | XP_011543298.1 | |||
| XM_011544997.2 | XP_011543299.1 | |||
| XM_011544998.2 | XP_011543300.1 | |||
| AK056195.1 | ||||
| AK307633.1 | ||||
| AK310714.1 | ||||
| BC098576.1 | AAH98576.1 | |||
| CV569756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751033 | Chr.11:62012550 - 62128252 on Build GRCh38 | Gain |
|
| esv2759832 | Chr.11:61981330 - 62126561 on Build GRCh38 | Gain |
|
| esv997129 | Chr.11:62123966 - 62125002 on Build GRCh38 | Gain |
|
| nsv1036085 | Chr.11:62104387 - 62127010 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141157640] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map