Assay Details
Target Gene Details
Entrez Gene ID: | 729025 |
Gene Name: | solute carrier family 15 member 5 |
Gene Aliases: |
- |
Location: |
Chr.12:16188485-16277685 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC15A5 | NM_001170798.1 | NP_001164269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036638 | Chr.12:15912446 - 16358900 on Build GRCh38 | Loss |
|
| esv3628686 | Chr.12:16217803 - 16220045 on Build GRCh38 | Loss |
|
| nsv1039831 | Chr.12:16119581 - 16239611 on Build GRCh38 | Loss |
|
| nsv832336 | Chr.12:16087099 - 16277470 on Build GRCh38 | Gain |
|
| esv3580162 | Chr.12:16216941 - 16239611 on Build GRCh38 | Loss |
|
| nsv428273 | Chr.12:16112158 - 16293444 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map