Assay Details
Target Gene Details
Entrez Gene ID: | 340527 |
Gene Name: | NHS like 2 |
Gene Aliases: |
- |
Location: |
Chr.X:71911073-72153286 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHSL2 | NM_001013627.2 | NP_001013649.2 | ||
| XM_011530932.1 | XP_011529234.1 | |||
| XM_011530933.1 | XP_011529235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758872 | Chr.X:71651403 - 72080566 on Build GRCh38 | Loss |
|
| nsv517419 | Chr.X:71888508 - 71924255 on Build GRCh38 | Gain |
|

Genomic Map