Assay Details
Target Gene Details
Entrez Gene ID: | 440590 |
Gene Name: | zyg-11 family member A, cell cycle regulator |
Gene Aliases: |
ZYG11 |
Location: |
Chr.1:52842511-52894575 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZYG11A | NM_001004339.2 | NP_001004339.2 | ||
| NM_001307931.1 | NP_001294860.1 | |||
| XM_011541480.2 | XP_011539782.1 | |||
| XM_011541481.2 | XP_011539783.1 | |||
| XM_011541482.2 | XP_011539784.1 | |||
| XM_011541483.2 | XP_011539785.1 | |||
| XM_011541484.1 | XP_011539786.1 | |||
| XM_011541485.1 | XP_011539787.1 | |||
| XM_011541486.1 | XP_011539788.1 | |||
| AY271826.1 | AAQ01753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| nsv829859 | Chr.1:52723470 - 52925273 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map