Assay Details
Target Gene Details
Entrez Gene ID: | 467 |
Gene Name: | activating transcription factor 3 |
Gene Aliases: |
- |
Location: |
Chr.1:212565334-212620777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF3 | NM_001030287.3 | NP_001025458.1 | ||
| AU135799.1 | ||||
| CD638624.1 | ||||
| CD641946.1 | ||||
| DA691940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549139 | Chr.1:212158952 - 212703560 on Build GRCh38 | Gain |
|
| esv2758996 | Chr.1:212474801 - 212669761 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79151334] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map