Assay Details
Target Gene Details
Entrez Gene ID: | 27246 |
Gene Name: | ring finger protein 115 |
Gene Aliases: |
BCA2, ZNF364 |
Location: |
Chr.1:145742189-145824123 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF115 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587421 | Chr.1:145649153 - 145751059 on Build GRCh38 | Loss |
|
| dgv331n100 | Chr.1:145642988 - 145827134 on Build GRCh38 | Gain |
|
| esv3572342 | Chr.1:145601946 - 145809476 on Build GRCh38 | Gain |
|
| nsv469752 | Chr.1:145639438 - 145790042 on Build GRCh38 | Loss |
|
| nsv831392 | Chr.1:145632374 - 145813165 on Build GRCh38 | Gain |
|
| esv3894089 | Chr.1:145672100 - 145824903 on Build GRCh38 | Loss |
|
| nsv547670 | Chr.1:145689573 - 145778504 on Build GRCh38 | Loss |
|
| dgv532n54 | Chr.1:145624561 - 145813526 on Build GRCh38 | Gain |
|
| dgv33n27 | Chr.1:145687602 - 145824903 on Build GRCh38 | Loss |
|
| dgv95e212 | Chr.1:145631218 - 145812561 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72997760] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map