Assay Details
Target Gene Details
Entrez Gene ID: | 5909 |
Gene Name: | RAP1 GTPase activating protein |
Gene Aliases: |
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP |
Location: |
Chr.1:21596215-21669444 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428432 | Chr.1:21409691 - 21615921 on Build GRCh38 | Gain |
|
| nsv834391 | Chr.1:21523440 - 21723989 on Build GRCh38 | Loss |
|
| esv34155 | Chr.1:21367084 - 21840123 on Build GRCh38 | Loss |
|
| nsv509002 | Chr.1:21578381 - 21654198 on Build GRCh38 | Insertion |
|
| nsv470704 | Chr.1:21590741 - 21611881 on Build GRCh38 | Gain |
|
| nsv954902 | Chr.1:21599708 - 21654507 on Build GRCh38 | Deletion |
|
| nsv517096 | Chr.1:21537198 - 21670355 on Build GRCh38 | Loss |
|
| nsv998678 | Chr.1:21564750 - 21614530 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map