Assay Details
Target Gene Details
Entrez Gene ID: | 10964 |
Gene Name: | interferon induced protein 44 like |
Gene Aliases: |
C1orf29, GS3686 |
Location: |
Chr.1:78620382-78646145 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFI44L | NM_006820.3 | 9 | 2912 | NP_006811.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3309841 | Chr.1:78643152 - 78643153 on Build GRCh38 | Mobile Element Insertion |
|
| esv3431836 | Chr.1:78643153 - 78643161 on Build GRCh38 | Insertion |
|
| nsv1001291 | Chr.1:78598959 - 79684864 on Build GRCh38 | Loss |
|
| esv3386975 | Chr.1:78643134 - 78643153 on Build GRCh38 | Insertion |
|
| esv3303563 | Chr.1:78643146 - 78643147 on Build GRCh38 | Mobile Element Insertion |
|
| esv3406252 | Chr.1:78643147 - 78643159 on Build GRCh38 | Insertion |
|
| nsv830315 | Chr.1:78542758 - 78734906 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map