Assay Details
Target Gene Details
Entrez Gene ID: | 100129620 |
Gene Name: | uncharacterized LOC100129620 |
Gene Aliases: |
- |
Location: |
Chr.1:99004276-99148852 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100129620 | NR_033940.1 | |||
| AK095544.1 | BAC04569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546884 | Chr.1:98987731 - 99062363 on Build GRCh38 | Loss |
|
| nsv830771 | Chr.1:98877965 - 99049970 on Build GRCh38 | Loss |
|
| nsv946090 | Chr.1:99007759 - 99008644 on Build GRCh38 | Duplication |
|

Genomic Map