Assay Details
Target Gene Details
Entrez Gene ID: | 100507670 |
Gene Name: | uncharacterized LOC100507670 |
Gene Aliases: |
- |
Location: |
Chr.1:151280024-151281929 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507670 | NR_135595.1 | |||
| BE385446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|

Genomic Map