Assay Details
Target Gene Details
Entrez Gene ID: | 100131564 |
Gene Name: | CCDC18 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:93310109-93345811 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC18-AS1 | NR_034089.1 | |||
| AL832786.1 |

Genomic Map