Assay Details
Target Gene Details
Entrez Gene ID: | 6125 |
Gene Name: | ribosomal protein L5 |
Gene Aliases: |
DBA6, L5, MSTP030, PPP1R135 |
Location: |
Chr.1:92832037-92841924 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL5 | NM_000969.3 | NP_000960.2 | ||
| AB055762.1 | BAB62856.1 | |||
| AB208980.1 | BAD92217.1 | |||
| AF113210.1 | AAG39281.1 | |||
| AK092963.1 | ||||
| AK095815.1 | ||||
| AK222604.1 | BAD96324.1 | |||
| AK314720.1 | ||||
| BC001882.1 | AAH01882.1 | |||
| BC109370.1 | AAI09371.1 | |||
| BC132970.1 | ||||
| BC132972.1 | ||||
| BG756697.1 | ||||
| BU941919.1 | ||||
| U14966.1 | AAA85654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508393 | Chr.1:92824956 - 92879367 on Build GRCh38 | Deletion |
|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
| nsv1008602 | Chr.1:92711254 - 93175251 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map