Assay Details
Target Gene Details
Entrez Gene ID: | 9887 |
Gene Name: | SMG7, nonsense mediated mRNA decay factor |
Gene Aliases: |
C1orf16, EST1C, SGA56M |
Location: |
Chr.1:183471962-183554193 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMG7 | XM_005245648.3 | XP_005245705.1 | ||
| XM_005245649.3 | XP_005245706.1 | |||
| XM_006711676.3 | XP_006711739.1 | |||
| XM_011510205.2 | XP_011508507.1 | |||
| XM_011510206.2 | XP_011508508.1 | |||
| XM_011510207.2 | XP_011508509.1 | |||
| XM_011510208.2 | XP_011508510.1 | |||
| XM_011510209.2 | XP_011508511.1 |
Target Gene Details
Entrez Gene ID: | 284649 |
Gene Name: | SMG7 antisense RNA 1 |
Gene Aliases: |
DKFZP564C196 |
Location: |
Chr.1:183460874-183471982 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMG7-AS1 | NR_040063.1 | |||
| AI793165.1 | ||||
| AL050020.1 | 1 | 294 |
More Information
Additional Information:
For this assay, SNP(s) [rs112259656] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map