Assay Details
Target Gene Details
Entrez Gene ID: | 54897 |
Gene Name: | castor zinc finger 1 |
Gene Aliases: |
CAS11, CST, SRG, ZNF693, dJ734G22.1 |
Location: |
Chr.1:10636602-10796676 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASZ1 | NM_001079843.2 | NP_001073312.1 | ||
| XM_005263479.3 | XP_005263536.1 | |||
| XM_017001539.1 | XP_016857028.1 | |||
| XM_017001540.1 | XP_016857029.1 | |||
| XM_017001541.1 | XP_016857030.1 | |||
| AA449398.1 | ||||
| DQ217660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4n27 | Chr.1:10613137 - 10736809 on Build GRCh38 | Loss |
|
| dgv105n54 | Chr.1:10613037 - 10736809 on Build GRCh38 | Loss |
|
| nsv828141 | Chr.1:10635420 - 10644140 on Build GRCh38 | Gain |
|
| esv29966 | Chr.1:10616743 - 10814095 on Build GRCh38 | Loss |
|
| nsv545400 | Chr.1:10624041 - 10781964 on Build GRCh38 | Loss |
|
| nsv525693 | Chr.1:10634068 - 10780848 on Build GRCh38 | Loss |
|
| nsv828030 | Chr.1:10617505 - 10735104 on Build GRCh38 | Gain |
|
| nsv953454 | Chr.1:10616244 - 10724643 on Build GRCh38 | Deletion |
|
| nsv828364 | Chr.1:10637753 - 10704107 on Build GRCh38 | Gain |
|
| nsv545401 | Chr.1:10630891 - 10697142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map