Assay Details
Target Gene Details
Entrez Gene ID: | 23443 |
Gene Name: | solute carrier family 35 member A3 |
Gene Aliases: |
AMRS |
Location: |
Chr.1:99969367-100026979 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35A3 | NM_001271684.1 | NP_001258613.1 | ||
| NM_001271685.1 | NP_001258614.1 | |||
| NM_012243.2 | NP_036375.1 | |||
| XM_005270691.4 | XP_005270748.1 | |||
| XM_011541135.2 | XP_011539437.1 | |||
| XM_011541136.2 | XP_011539438.1 | |||
| XM_011541137.2 | XP_011539439.1 | |||
| XM_011541138.2 | XP_011539440.1 | |||
| XM_017000869.1 | XP_016856358.1 | |||
| XM_017000870.1 | XP_016856359.1 | |||
| XM_017000871.1 | XP_016856360.1 | |||
| XM_017000872.1 | XP_016856361.1 | |||
| AB021981.1 | BAA77841.1 | |||
| AK024989.1 | ||||
| AK290573.1 | ||||
| AV698459.1 | ||||
| BC005136.2 | AAH05136.1 | |||
| CR749816.1 | CAH18676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758952 | Chr.1:99894099 - 100032786 on Build GRCh38 | Loss |
|
| nsv546888 | Chr.1:99954311 - 100084889 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map