Assay Details
Target Gene Details
Entrez Gene ID: | 84144 |
Gene Name: | synapse defective Rho GTPase homolog 2 |
Gene Aliases: |
- |
Location: |
Chr.1:85157673-85201725 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYDE2 | NM_032184.1 | 2 | 1144 | NP_115560.1 |
| XM_005271254.3 | 2 | 1822 | XP_005271311.1 | |
| XM_006710946.3 | 2 | 1823 | XP_006711009.1 | |
| XM_017002483.1 | 2 | 1824 | XP_016857972.1 | |
| XM_017002484.1 | 2 | 1823 | XP_016857973.1 | |
| XM_017002485.1 | 2 | 531 | XP_016857974.1 | |
| XM_017002486.1 | 2 | 444 | XP_016857975.1 | |
| AL834286.1 | 2 | 612 | CAD38960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34165 | Chr.1:85076357 - 85380566 on Build GRCh38 | Loss |
|
| nsv830459 | Chr.1:85123902 - 85320628 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map