Assay Details
Target Gene Details
Entrez Gene ID: | 22926 |
Gene Name: | activating transcription factor 6 |
Gene Aliases: |
ACHM7, ATF6A |
Location: |
Chr.1:161766244-161964070 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF6 | NM_007348.3 | NP_031374.2 | ||
| XM_006711224.1 | XP_006711287.1 | |||
| XM_011509308.1 | XP_011507610.1 | |||
| XM_011509309.1 | XP_011507611.1 | |||
| XM_011509310.2 | XP_011507612.1 | |||
| AB015856.1 | BAA34722.1 | |||
| AB208929.1 | BAD92166.1 | |||
| AF005887.1 | AAB64434.1 | |||
| AK290498.1 | ||||
| BC014969.1 | AAH14969.1 | |||
| BC071997.1 | AAH71997.1 | |||
| BP380917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv548075 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map