Assay Details
Target Gene Details
Entrez Gene ID: | 9909 |
Gene Name: | DENN domain containing 4B |
Gene Aliases: |
KIAA0476 |
Location: |
Chr.1:153929501-153946681 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND4B | NM_014856.2 | NP_055671.2 | ||
| XM_005245678.4 | XP_005245735.1 | |||
| XM_005245679.2 | XP_005245736.1 | |||
| XM_011510219.1 | XP_011508521.1 | |||
| XM_011510220.2 | XP_011508522.1 | |||
| XM_011510221.2 | XP_011508523.1 | |||
| XM_011510222.1 | XP_011508524.1 | |||
| XM_017002993.1 | XP_016858482.1 | |||
| AB007945.2 | ||||
| AK129713.1 | 1 | 1025 | ||
| AK226053.1 | ||||
| L10375.1 |
More Information
Set Membership: |
|

Genomic Map