Assay Details
Target Gene Details
Entrez Gene ID: | 462 |
Gene Name: | serpin family C member 1 |
Gene Aliases: |
AT3, AT3D, ATIII, THPH7 |
Location: |
Chr.1:173903804-173917378 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINC1 | NM_000488.3 | NP_000479.1 | ||
| XM_005245198.3 | XP_005245255.1 | |||
| AF130100.1 | AAG35525.1 | |||
| AK312654.1 | ||||
| BC022309.1 | AAH22309.1 | |||
| D29832.1 | BAA06212.1 | |||
| KP780080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758978 | Chr.1:173841504 - 173986805 on Build GRCh38 | Loss |
|
| nsv831926 | Chr.1:173733264 - 173925529 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map