Assay Details
Target Gene Details
Entrez Gene ID: | 10964 |
Gene Name: | interferon induced protein 44 like |
Gene Aliases: |
C1orf29, GS3686 |
Location: |
Chr.1:78620382-78646145 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFI44L | NM_006820.3 | NP_006811.2 | ||
| XM_005270391.3 | XP_005270448.1 | |||
| XM_005270392.4 | XP_005270449.1 | |||
| XM_005270393.4 | XP_005270450.1 | |||
| XM_006710304.3 | XP_006710367.1 | |||
| XM_011540539.2 | XP_011538841.1 | |||
| XM_017000120.1 | XP_016855609.1 | |||
| AB000115.1 | BAA19056.1 | |||
| AK223087.1 | BAD96807.1 | |||
| AK225442.1 | ||||
| AK293385.1 | ||||
| AK303187.1 | ||||
| AK312272.1 | ||||
| AK316232.1 | ||||
| AL832618.2 | CAD90004.1 | |||
| BC015932.2 | AAH15932.1 | |||
| DC390321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2661424 | Chr.1:78628775 - 78631915 on Build GRCh38 | Deletion |
|
| nsv1001291 | Chr.1:78598959 - 79684864 on Build GRCh38 | Loss |
|
| esv3586552 | Chr.1:78628933 - 78631885 on Build GRCh38 | Loss |
|
| nsv830315 | Chr.1:78542758 - 78734906 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map