Assay Details
Target Gene Details
Entrez Gene ID: | 728621 |
Gene Name: | coiled-coil domain containing 30 |
Gene Aliases: |
PFD6L, PFDN6L |
Location: |
Chr.1:42521240-42654664 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC30 | NM_001080850.2 | 2 | 183 | NP_001074319.1 |
| XM_011542067.2 | 3 | 1115 | XP_011540369.1 | |
| XM_011542068.2 | 3 | 1120 | XP_011540370.1 | |
| XM_011542069.2 | 3 | 1112 | XP_011540371.1 | |
| XM_011542071.2 | 3 | 1108 | XP_011540373.1 | |
| XM_017002225.1 | 2 | 200 | XP_016857714.1 | |
| XM_017002226.1 | 3 | 511 | XP_016857715.1 | |
| AK024625.1 | 6 | 618 | BAB14936.1 | |
| AK302075.1 | 5 | 505 | ||
| AL832217.1 | 9 | 1216 | ||
| AY639646.1 | ||||
| BX640642.1 | CAE45789.1 | |||
| DC397297.1 |

Genomic Map