Assay Details
Target Gene Details
Entrez Gene ID: | 100131564 |
Gene Name: | CCDC18 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:93310109-93345811 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC18-AS1 | NR_034089.1 | |||
| AL832786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586828 | Chr.1:93338495 - 93341568 on Build GRCh38 | Loss |
|

Genomic Map