Assay Details
Target Gene Details
Entrez Gene ID: | 57643 |
Gene Name: | zinc finger SWIM-type containing 5 |
Gene Aliases: |
- |
Location: |
Chr.1:45016399-45206586 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSWIM5 | NM_020883.1 | NP_065934.1 | ||
| XM_011541861.2 | XP_011540163.1 | |||
| XM_017001913.1 | XP_016857402.1 | |||
| AB040944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546148 | Chr.1:45034622 - 45161991 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map