Assay Details
Target Gene Details
| Entrez Gene ID: | 54802 | 
| Gene Name: | tRNA isopentenyltransferase 1 | 
| Gene Aliases: | GRO1, IPPT, IPT, IPTase, MOD5, hGRO1 | 
| Location: | Chr.1:39841031-39883511 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |  MYCL  BMP8B  LOC101929536  RLF  OXCT2  PPIE  TRIT1  CAP1  PPT1  MFSD2A | 
| esv2666580 | Chr.1:39809257 - 39898016 on Build GRCh38 | Deletion |  MYCL  TRIT1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top