Assay Details
Target Gene Details
Entrez Gene ID: | 23418 |
Gene Name: | crumbs 1, cell polarity complex component |
Gene Aliases: |
LCA8, RP12 |
Location: |
Chr.1:197201462-197478455 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRB1 | NM_001257965.1 | NP_001244894.1 | ||
| AK302826.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588493 | Chr.1:197157545 - 197347526 on Build GRCh38 | Loss |
|
| nsv528460 | Chr.1:196573437 - 197477572 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map