Assay Details
Target Gene Details
Entrez Gene ID: | 105376736 |
Gene Name: | uncharacterized LOC105376736 |
Gene Aliases: |
- |
Location: |
Chr.1:11099675-11102105 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376736 | NR_135061.1 | 2 | 467 | |
| NR_135062.1 | 2 | 371 | ||
| NR_135063.1 | 2 | 297 | ||
| AI342698.1 | 2 | 371 | ||
| AI460252.1 | ||||
| BE669601.1 | ||||
| BG184665.1 | 1 | 239 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|

Genomic Map