Assay Details
Target Gene Details
Entrez Gene ID: | 91408 |
Gene Name: | basic transcription factor 3 like 4 |
Gene Aliases: |
- |
Location: |
Chr.1:52056185-52090716 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTF3L4 | NM_001136497.2 | NP_001129969.1 | ||
| NM_001243767.1 | NP_001230696.1 | |||
| NM_152265.4 | NP_689478.1 | |||
| AK027582.1 | ||||
| AK027750.1 | BAB55342.1 | |||
| BC021004.1 | AAH21004.1 | |||
| BC022371.1 | AAH22371.1 | |||
| BC070378.1 | AAH70378.1 | |||
| BU567198.1 | ||||
| BU785869.1 | ||||
| DA369498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004103 | Chr.1:52030594 - 52087736 on Build GRCh38 | Loss |
|
| nsv829848 | Chr.1:51902939 - 52076027 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map