Assay Details
Target Gene Details
Entrez Gene ID: | 7062 |
Gene Name: | trichohyalin |
Gene Aliases: |
THH, THL, TRHY |
Location: |
Chr.1:152106317-152115454 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCHH | NM_007113.3 | 2 | 131 | NP_009044.2 |
| AY827491.1 | 2 | 121 | AAX12418.1 | |
| BC131826.1 | 1 | 1495 | ||
| BC131827.1 | 1 | 1495 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
| nsv2866 | Chr.1:152088370 - 152120918 on Build GRCh38 | Insertion |
|
| nsv547853 | Chr.1:152036195 - 152264284 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map