Assay Details
Target Gene Details
Entrez Gene ID: | 261734 |
Gene Name: | nephronophthisis 4 |
Gene Aliases: |
POC10, SLSN4 |
Location: |
Chr.1:5862808-5992473 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545173 | Chr.1:5833391 - 6025293 on Build GRCh38 | Gain |
|
| nsv508723 | Chr.1:5777215 - 5971948 on Build GRCh38 | Insertion |
|
| nsv545174 | Chr.1:5844658 - 5971884 on Build GRCh38 | Loss |
|
| nsv831203 | Chr.1:5830030 - 5967902 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map