Assay Details
Target Gene Details
Entrez Gene ID: | 83998 |
Gene Name: | regenerating family member 4 |
Gene Aliases: |
GISP, REG-IV, RELP |
Location: |
Chr.1:119794018-119811580 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REG4 | NM_001159352.1 | NP_001152824.1 | ||
| NM_001159353.1 | NP_001152825.1 | |||
| NM_032044.3 | NP_114433.1 | |||
| AF254415.1 | AAK48435.1 | |||
| AF345934.1 | AAK59869.1 | |||
| AY007243.1 | AAG02562.1 | |||
| AY126670.1 | AAM95598.1 | |||
| AY126671.1 | AAM95599.1 | |||
| AY126672.1 | AAM95600.1 | |||
| BC017089.2 | AAH17089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831226 | Chr.1:119788976 - 119980845 on Build GRCh38 | Gain+Loss |
|
| nsv831215 | Chr.1:119723826 - 119905846 on Build GRCh38 | Gain |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
| nsv428201 | Chr.1:119724690 - 120154589 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map