Assay Details
Target Gene Details
Entrez Gene ID: | 249 |
Gene Name: | alkaline phosphatase, liver/bone/kidney |
Gene Aliases: |
AP-TNAP, APTNAP, HOPS, TNAP, TNSALP |
Location: |
Chr.1:21508982-21578412 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALPL | NM_000478.5 | NP_000469.3 | ||
| NM_001127501.3 | NP_001120973.2 | |||
| NM_001177520.2 | NP_001170991.1 | |||
| XM_005245818.1 | XP_005245875.1 | |||
| AB011406.1 | BAA32129.1 | |||
| AB209814.1 | BAD93051.1 | |||
| AK293184.1 | ||||
| AK295608.1 | ||||
| AK298085.1 | ||||
| AK312667.1 | ||||
| BC021289.2 | ||||
| BC066116.1 | ||||
| BC090861.1 | ||||
| BC110909.1 | ||||
| CN425768.1 | ||||
| DA574668.1 | ||||
| DA631560.1 | ||||
| HY052024.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428432 | Chr.1:21409691 - 21615921 on Build GRCh38 | Gain |
|
| esv34155 | Chr.1:21367084 - 21840123 on Build GRCh38 | Loss |
|
| dgv24e212 | Chr.1:21516950 - 21524296 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116905590] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map