Assay Details
Target Gene Details
Entrez Gene ID: | 149111 |
Gene Name: | cornichon family AMPA receptor auxiliary protein 3 |
Gene Aliases: |
CNIH-3 |
Location: |
Chr.1:224434640-224740549 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNIH3 | NM_001322302.1 | NP_001309231.1 | ||
| NM_001322303.1 | NP_001309232.1 | |||
| NM_001322304.1 | NP_001309233.1 | |||
| NR_136288.1 | ||||
| NR_136289.1 | ||||
| NR_136290.1 | ||||
| NR_136291.1 | ||||
| NR_136292.1 | ||||
| NR_136293.1 | ||||
| NR_136294.1 | ||||
| NR_136295.1 | ||||
| NR_136296.1 | ||||
| NR_136297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8868 | Chr.1:224195321 - 224564829 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map