Assay Details
Target Gene Details
Entrez Gene ID: | 126767 |
Gene Name: | arylacetamide deacetylase like 3 |
Gene Aliases: |
- |
Location: |
Chr.1:12716115-12728759 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL3 | NM_001103170.2 | 4 | 1221 | NP_001096640.2 |
| NR_111984.1 | 3 | 1004 | ||
| AK127828.1 | 3 | 1004 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011540 | Chr.1:12601623 - 12832443 on Build GRCh38 | Loss |
|
| nsv545445 | Chr.1:12714996 - 12854043 on Build GRCh38 | Gain |
|
| nsv545444 | Chr.1:12687857 - 12727665 on Build GRCh38 | Loss |
|
| nsv999658 | Chr.1:12708960 - 12799252 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map