Assay Details
Target Gene Details
Entrez Gene ID: | 728621 |
Gene Name: | coiled-coil domain containing 30 |
Gene Aliases: |
PFD6L, PFDN6L |
Location: |
Chr.1:42521240-42654664 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC30 | XM_011542067.2 | XP_011540369.1 | ||
| XM_011542068.2 | XP_011540370.1 | |||
| XM_011542069.2 | XP_011540371.1 | |||
| XM_011542071.2 | XP_011540373.1 | |||
| XM_017002226.1 | XP_016857715.1 | |||
| AK024625.1 | BAB14936.1 | |||
| AK302075.1 | ||||
| AL832217.1 | ||||
| AY639646.1 | ||||
| DC397297.1 |

Genomic Map